Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PICALM Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$682.00
Specifications
Antigen | PICALM |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PICALM Polyclonal specifically detects PICALM in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PICALM | |
Polyclonal | |
Rabbit | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
8301 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DIPDLSQAPSSLLDALEQHLASLEGKKIKDSTAASRATTLSNAVSSLASTGLSLTKVDEREKQAALEEEQARLKALKEQRL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
CALMclathrin assembly lymphoid-myeloid leukemia, Clathrin assembly lymphoid myeloid leukemia protein, CLTHphosphatidylinositol-binding clathrin assembly protein, LAP, phosphatidylinositol binding clathrin assembly protein | |
PICALM | |
IgG | |
Affinity Purified | |
Specificity of human PICALM antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title