Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Piccolo Antibody (6H9-B6), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $560.50
Specifications
Antigen | Piccolo |
---|---|
Clone | 6H9-B6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:2000, ELISA 1 ug/ml, Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP149453SS
![]() |
Novus Biologicals
NBP149453SS |
0.025 mg |
Each for $268.00
|
|
|||||
NBP149453
![]() |
Novus Biologicals
NBP149453 |
0.1 mg |
Each for $560.50
|
|
|||||
Description
Piccolo Monoclonal specifically detects Piccolo in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Piccolo | |
1 mg/mL | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
piccolo (presynaptic cytomatrix protein), protein piccolo | |
PCLO | |
IgG2b κ | |
Protein G purified | |
Detects approx 550kDa. |
6H9-B6 | |
Western Blot 1:2000, ELISA 1 ug/ml, Immunocytochemistry/Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
BC001304.1 | |
27445 | |
Full length protein, BC001304.1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
550 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title