Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Piccolo Antibody (PCLO-01), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262209AF532
Description
Piccolo Monoclonal specifically detects Piccolo in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
| Piccolo | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PCLO | |
| Human recombinant PCLO protein | |
| 0.1 mL | |
| 27445 | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| PCLO-01 | |
| Western Blot, Flow Cytometry | |
| piccolo (presynaptic cytomatrix protein), protein piccolo | |
| Mouse | |
| Protein A purified | |
| Primary | |
| The antibody A53-B/A2 reacts with Rod domain of cytokeratin 19 (40 kDa) in human tissue. Cytokeratin 19 is not expressed in hepatocytes; it is often co-expressed with cytokeratin 7. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction