Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Piccolo Antibody (PCLO-01), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262209AF700
Description
Piccolo Monoclonal specifically detects Piccolo in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
Piccolo | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PCLO | |
Human recombinant PCLO protein | |
0.1 mL | |
27445 | |
Human | |
IgG1 |
Western Blot, Flow Cytometry | |
PCLO-01 | |
Western Blot, Flow Cytometry | |
piccolo (presynaptic cytomatrix protein), protein piccolo | |
Mouse | |
Protein A purified | |
Primary | |
The antibody A53-B/A2 reacts with Rod domain of cytokeratin 19 (40 kDa) in human tissue. Cytokeratin 19 is not expressed in hepatocytes; it is often co-expressed with cytokeratin 7. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction