Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piccolo Monoclonal Antibody (6H9-B6), QED Bioscience™
SDP

Catalog No. 502278202
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8202 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8202 Supplier Leinco Technologies Inc Supplier No. 56484100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Piccolo Monoclonal antibody specifically detects Piccolo in Human, Rat samples. It is validated for ELISA, Western Blot

Specifications

Antigen Piccolo
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6H9-B6
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene PCLO
Gene Accession No. Q9JKS6, Q9Y6V0
Gene Alias Acz; Aczonin; brain-derived HLMN protein; KIAA0559; LOW QUALITY PROTEIN: protein piccolo; multidomain presynaptic cytomatrix protein; multidomain presynaptic cytomatrix protein Piccolo; PCH3; PCLO; piccolo (presynaptic cytomatrix protein); piccolo presynaptic cytomatrix protein; Pico; presynaptic cytomatrix protein; protein piccolo
Host Species Mouse
Immunogen Full-length human Piccolo protein (accession no. BC001304.1)
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27445, 56768
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.