Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piceatannol 98.0+%, TCI America™
SDP

Catalog No. p-7135319
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 10083-24-6
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00221715
InChI Key CDRPUGZCRXZLFL-OWOJBTEDSA-N
Synonym piceatannol, 3-hydroxyresveratol, astringinin, piceatanol, e-4-3,5-dihydroxystyryl benzene-1,2-diol, 3,3',4'5-tetrahydroxystilbene, 3,5,3',4'-tetrahydroxystilbene, 4-e-2-3,5-dihydroxyphenyl ethenyl benzene-1,2-diol, 3,3',4,5'-tetrahydroxystilbene, unii-6ks3ls0d4f
PubChem CID 667639
ChEBI CHEBI:28814
IUPAC Name 4-[(E)-2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,2-diol
SMILES C1=CC(=C(C=C1C=CC2=CC(=CC(=C2)O)O)O)O

Specifications

Melting Point 235°C
Color Red-White
Boiling Point 108°C
Quantity 100 mg
Formula Weight 244.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Piceatannol
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.