Learn More
Invitrogen™ Pichia pastoris Polyclonal Antibody

Description
PA1-7237 detects the Pichia pastoris. This antibody cross reacts with Candida albicans (strong) and S. cerevisiae (weak). PA1-7237 has been successfully used in immunofluorescence, and Western blot procedures. The PA1-7237 immunogen is whole cells of Pichia pastoris.
Specifications
Specifications
| Antigen | Pichia pastoris |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | P. pastoris |
| Host Species | Rabbit |
| Immunogen | Whole cells of Pichia pastoris. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.