Learn More
Invitrogen™ PICK1 Polyclonal Antibody

Description
Immunogen sequence: LTIKKYLDVKF EYLSYCLKVK EMDDEEYSCI ALGEPLYRVS TGNYEYRLIL RCRQEARARF SQMRKDVLEK MELLD Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Specifications
Specifications
| Antigen | PICK1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Pick1 |
| Gene Accession No. | Q9NRD5 |
| Gene Alias | MGC15204; OTTHUMP00000028509; PICK; Pick1; PRKCA-binding protein; Prkcabp; Protein interacting with C kinase 1; protein interacting with PRKCA 1; protein kinase C, alpha binding protein; protein kinase C-alpha-binding protein; protein that interacts with C kinase 1 |
| Gene Symbols | Pick1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.