Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PICK1 Polyclonal Antibody, Invitrogen™

Catalog No. PA1073
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1073 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1073 Supplier Thermo Scientific Supplier No. PA1073

Rabbit Polyclonal Antibody

PA1-073 detects protein interacting with protein kinase C (PICK1) from mouse cells. PA1-073 has been successfully used in Western blot, immunocytochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects an ∽52 kDa protein representing PICK1 from AtT20 cell extract. Immunocytochemical staining of CV1 cells transfected with PICK1 results in a diffuse staining pattern when expressed alone and coclustered when coexpressed with mGluR7. The PA1-073 immunogen is a synthetic peptide corresponding to residues M(1) F A D L D Y D I E E D K L G I P T(18) of mouse PICK1. The PA1-073 immunizing peptide (Cat. # PEP-133) is available for use in neutralization and control experiments.

PSD-95/DLG/ZO-1 (PDZ) domain-containing proteins play a central role in synaptic membrane protein localization. The protein interacting with protein kinase C (PICK1) is a synaptic PDZ domain protein that also contains a coiled-coil and acidic domain. Studies indicate that PICK1 functions as a targeting and transport protein and has been shown to interact with protein kinase C alpha (PKCalpha), AMPA-type glutamate receptors, and several other membrane receptors via its PDZ domain. The interaction of PICK1 with PKCalpha is highly dependent on the activation of the kinase. It appears that PICK1 directs the activated form of PKCalpha to membrane anchored GluR2. Once phosphorylated by PKCalpha, GluR2 is released from the synaptic anchor proteins. Once released, GluR2 is transported from the synaptic membrane in a PICK1-dependent manner.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

TRUSTED_SUSTAINABILITY

Specifications

Antigen PICK1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Pick1
Gene Accession No. Q62083, Q9EP80
Gene Alias MGC15204; OTTHUMP00000028509; PICK; Pick1; PRKCA-binding protein; Prkcabp; Protein interacting with C kinase 1; protein interacting with PRKCA 1; protein kinase C, alpha binding protein; protein kinase C-alpha-binding protein; protein that interacts with C kinase 1
Gene Symbols Pick1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) F A D L D Y D I E E D K L G I P T(18) of mouse PICK1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18693, 84591
Target Species Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.