Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PICK1 Recombinant Rabbit Monoclonal Antibody (JG30-80)
GREENER_CHOICE

Catalog No. PIMA534772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534772 Supplier Invitrogen™ Supplier No. MA534772
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: SK-Br-3, 293T.

PSD-95/DLG/ZO-1 (PDZ) domain-containing proteins play a central role in synaptic membrane protein localization. The protein interacting with protein kinase C (PICK1) is a synaptic PDZ domain protein that also contains a coiled-coil and acidic domain. Studies indicate that PICK1 functions as a targeting and transport protein and has been shown to interact with protein kinase C alpha (PKCalpha), AMPA-type glutamate receptors, and several other membrane receptors via its PDZ domain. The interaction of PICK1 with PKCalpha is highly dependent on the activation of the kinase. It appears that PICK1 directs the activated form of PKCalpha to membrane anchored GluR2. Once phosphorylated by PKCalpha, GluR2 is released from the synaptic anchor proteins. Once released, GluR2 is transported from the synaptic membrane in a PICK1-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PICK1
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone JG30-80
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Pick1
Gene Accession No. Q9NRD5
Gene Alias MGC15204; OTTHUMP00000028509; PICK; Pick1; PRKCA-binding protein; Prkcabp; Protein interacting with C kinase 1; protein interacting with PRKCA 1; protein kinase C, alpha binding protein; protein kinase C-alpha-binding protein; protein that interacts with C kinase 1
Gene Symbols Pick1
Host Species Rabbit
Immunogen Synthetic peptide within Human PICK1 aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9463
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.