Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Piclamilast
GREENER_CHOICE

Catalog No. 452510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-251-0 10 mg
45-255-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-251-0 Supplier Tocris Bioscience™ Supplier No. 4525/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PDE4 inhibitor

Potent and selective inhibitor of phosphodiesterase (PDE) 4 (IC50 = 1 nM at PDE4 from human neutrophils). Displays >19,000-fold selectivity over other PDE isoenzymes. Exhibits anti-inflammatory effects in models of allergic inflammation. Also inhibits LTB4 synthesis in human neutrophils (IC50 = 2 nM).

Chemical Identifiers

CAS 144035-83-6
Molecular Formula C18H18Cl2N2O3

Specifications

Quantity 10 mg
Formula Weight 381.25
Purity >99%
Molecular Weight (g/mol) 381.253
Chemical Name or Material Piclamilast
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.