Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picloram, 95%

Catalog No. AAJ6670206
Click to view available options
Quantity:
5 g
25 g

Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA) | CAS: 1918-02-1 | C6H3Cl3N2O2 | 241.45 g/mol

Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1918-02-1
Molecular Formula C6H3Cl3N2O2
Molecular Weight (g/mol) 241.45
MDL Number MFCD00012101
InChI Key NQQVFXUMIDALNH-UHFFFAOYSA-N
Synonym picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k
PubChem CID 15965
ChEBI CHEBI:34922
IUPAC Name 4-amino-3,5,6-trichloropyridine-2-carboxylic acid
SMILES C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N

Specifications

Melting Point 219°C
Color White
Boiling Point 421°C
Flash Point >110°C (230°F)
Odor Chlorine-like
Quantity 5 g
Beilstein 479075
Merck Index 14,7397
Solubility Information Soluble in acetone
Formula Weight 241.46
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Picloram
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 217-636-1
RTECSNumber TJ7525000
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.