Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picric Acid, Crystal, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18605215
Click to view available options
Quantity:
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Drum

P1145, 88-89-1, C6H2(NO2)3OH

Spectrum™ Chemical Picric Acid, Crystal, Reagent, ACS , also known as 2,4,6-trinitrophenol or TNP. It is a yellow crystalline solid and one of the most acidic phenols. As a highly nitrated compound, picric acid is highly explosive (like TNT). Apart from it's use as an explosive, it also has some uses in organic chemistry for the preparation of crystalline salts of organic bases. It is also used in clinical chemistry for the Jaffe reaction (a colorimetric method to determine creatinine levels in blood and urine).

Chemical Identifiers

CAS 88-89-1
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) 229.10
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
IUPAC Name 2,4,6-trinitrophenol
SMILES OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Max % 1
Grade ACS
Quantity 12 kg
Packaging Poly Drum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.