Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picric Acid, Saturated (AOAC), Ricca Chemical
SDP

Catalog No. sp920050 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 7732-18-5, 88-89-1
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) Mixture
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
Synonym picric acid, trinitrophenol, carbazotic acid, picronitric acid, melinite, acide picrique, phenol trinitrate, picral, nitroxanthic acid, pikrinsaeure
PubChem CID 6954
ChEBI CHEBI:46149
IUPAC Name 2,4,6-trinitrophenol
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

CAS Min % 97.0, 1.26
CAS Max % 100.0, 1.31
Grade Laboratory
Quantity 50 mL
Packaging Natural Poly Bottle
Chemical Name or Material Picric Acid, Saturated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.