Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Picropodophyllotoxin
GREENER_CHOICESDP

Catalog No. 295610 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-561-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-561-0 Supplier R&D Systems™ Supplier No. 2956/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective IGF1R inhibitor

Orally active insulin-like growth factor 1 receptor (IGF1R) inhibitor that exhibits no activity at the insulin receptor, FGFR, PDGFR or EGFR. Inhibits IGF1R autophosphorylation (IC50 ∽ 1 nM), increases the fraction of cells in the G2/M phase and upregulates apoptosis. Exhibits antiproliferative effects in multiple cancer cell lines (IC50 = 0.05 - 15 μM), and has anticancer and antineovascularization activity in vivo.

Chemical Identifiers

CAS 477-47-4
Molecular Formula C22H22O8

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 414.41
Purity >97%
Molecular Weight (g/mol) 414.41
Chemical Name or Material Picropodophyllotoxin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.