Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIDD Antibody (Lise-1), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311673V
Description
LRDD Monoclonal specifically detects LRDD in Mouse samples. It is validated for Western Blot.Specifications
LRDD | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Lise-1 | |
Western Blot | |
leucine-rich repeat and death domain-containing protein, leucine-rich repeats and death domain containing, MGC16925, p53-induced protein with a death domain, PIDD | |
Mouse MDA5 (Death Domain). | |
0.1 mL | |
Cell Cycle and Replication | |
55367 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction