Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIDD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544740 Supplier Invitrogen™ Supplier No. PA544740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RLQGNPLGEA SPDAPSSPVA ALIPEMPRLF LTSDLDSFPV TPQGCSVTLA Sequence homology: Human: 100%; Mouse: 86%; Rat: 100%.

The protein encoded by this gene contains a leucine-rich repeat and a death domain. This protein has been shown to interact with other death domain proteins, such as Fas (TNFRSF6)-associated via death domain (FADD) and MAP-kinase activating death domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIDD
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PIDD1
Gene Accession No. Q9HB75
Gene Alias 1200011D09Rik; AU042446; DKFZp434D229; leucine-rich and death domain containing; leucine-rich repeat and death domain-containing protein; leucine-rich repeats and death domain containing; Lrdd; MGC16925; p53 induced death domain protein 1; p53 protein induced, with death domain; p53-induced death domain protein 1; p53-induced death domain-containing protein 1; p53-induced protein with a death domain; Pidd; Pidd1; PIDD-C; PIDD-CC; PIDD-N
Gene Symbols PIDD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LRDD (aa 289-338).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55367
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.