Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pidotimod 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
Specifications
Specifications
Purity | ≥98.0% (HPLC,T) |
CAS | 121808-62-6 |
Color | White |
Format | Crystal-Powder at 20°C |
Molecular Weight (g/mol) | 244.27 |
Product Type | Pidotimod |
Quantity | 5g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction