Learn More
Thermo Scientific™ 0.1% Formic Acid (v/v) in Acetonitrile, LC-MS Grade
Description
Thermo Scientific™ 0.1% Formic Acid in Acetonitrile is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
Highlights:
Validated – tested more than 20 ways to ensure maximal LC/MS sensitivity
Purified – processed, filtered and sealed to extend LC/MS column life
Convenient – packaged to eliminate variability and reduce handling
Specifications
Specifications
| Grade | LC-MS |
| Item Name | Formic Acid |
| Quantity | 4 x 1 L |
| Content And Storage | Store in a cool, dry, and well-ventilated area. Protect from direct sunlight. |
| Form | Liquid |
| Reagent Type | Solvent Mixture |
| Product Line | Pierce |
| For Use With | HPLC, Mass Spectrometry |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.