Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 0.1% Trifluoroacetic Acid (v/v) in Water, LC-MS Grade

Catalog No. PI85173
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI85173 4 x 1 L
PI85172 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI85173 Supplier Thermo Scientific™ Supplier No. 85173

An LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

Thermo Scientific™ 0.1% Trifluoroacetic Acid in Water is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

This 0.1% Trifluoroacetic Acid in Water is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography-and spectrometry-grade 0.1% TFA in water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Validated

    – tested more than 20 ways to ensure maximal LC/MS sensitivity

  • Purified

    – processed, filtered and sealed to extend LC/MS column life

  • Convenient

    – packaged to eliminate variability and reduce handling

TRUSTED_SUSTAINABILITY

Specifications

Grade LC-MS
Item Name Trifluoroacetic Acid (TFA)
Quantity 4 x 1 L
Content And Storage Store in a cool, dry, and well-ventilated area. Protect from direct sunlight.
Form Liquid
Reagent Type Solvent Mixture, Ion Pairing Reagent
Product Line Pierce
For Use With HPLC, Mass Spectrometry
What kind of buffers and solutions are compatible with peptide clean-up and LC-MS?

Most organic solvents or volatile buffers are compatible with LC-MS. Water and acetonitrile with 0.1% formic acid is the most common solvent system for separation of peptide samples for mass spectrometry analysis. Trifluoracetic acid (TFA) is an alternative ion pairing agent for off-line peptide C18 clean-up or LC-UV analysis. We offer the following LC-MS grade reagents: Pierce 0.1% Trifluoroacetic Acid (v/v) in Water (Cat. No. 85172), Pierce Water, LC-MS Grade (Cat. No. 85189), Pierce Acetonitrile (ACN) (Cat. No. 85188), and Pierce Trifluoroacetic (TFA) (Cat. No. 85183).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.