Learn More
Thermo Scientific™ Pierce™ 6xHis Protein Tag Stain Reagent Set
Description
Thermo Scientific™ Pierce 6xHis Protein Tag Stain Kit is a fluorescent stain for preferential detection of histidine-tagged (His-tagged) recombinant fusion proteins in protein polyacrylamide gels.
Highlights:
Two- to three-times faster than Western blotting – get results sooner and save hours of valuable research time
Detects directly on the gel – can eliminate the need for a membrane transfer and Western blot step for the detection of at least 0.2μg of 6xHis-tagged protein
Ready-to-use, two-reagent formula – no mixing, no diluting and no fuss; guarantees a simple-to-perform, mistake-free detection protocol
Specific fluorescent detection of 6xHis-tagged proteins – see only what you want to see (CCD camera recommended for low abundance proteins; UV-transilluminator for abundant proteins)
Compatible with double-staining – follow His-tagged protein staining with Thermo Scientific GelCode Blue Stain Reagent for a total protein profile determination

Specifications
Specifications
Content And Storage | Store components at room temperature. Product is shipped at ambient temperature |
Description | Pierce 6xHis Protein Tag Stain Reagent Set |
Detection Location | In-Gel Detection |
Detection Method | Fluorescence |
Label or Dye | Proprietary Mix |
Product Type | His Tag Protein Stain Set |
Product Line | Pierce™ |
Quantity | 1 L |
Target Molecule | Proteins (His-Tagged) |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.