Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ Alkaline Phosphatase
Click to view available options
Quantity:
20 mg
5 x 20 mg
Description
Thermo Scientific™ Pierce Alkaline Phosphatase is purified calf-intestinal alkaline phosphatase (AP or alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and immunohistochemistry applications.
Highlights:
- Purified form - ready to conjugate without prior dialysis
- Activity is not affected by exposure to antibacterial agents, such as sodium azide or thimerosal
- Specific activity > 2,000 units/mg
- One unit is defined as the amount that will hydrolyze 1.0 mol of p-nitrophenyl phosphate per minute at 37° C in 1.0M diethanolamine, 0.5mM MgCl2, pH 7.8
Specifications
Specifications
| Content And Storage | Store at 2–8°C. |
| Form | Solid |
| For Use With (Application) | ELISA, Immunohistochemistry |
| Product Line | Pierce |
| Label or Dye | AP (Alkaline Phosphatase) |
| Quantity | 20 mg |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction