Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ Anti-DYKDDDDK Magnetic Agarose, 4 mL PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ Anti-DYKDDDDK Magnetic Agarose
Description
- Thermo Scientific Pierce Anti-DYKDDDDK Magnetic Agarose provides a fast, convenient method for purification and immunoprecipitation (IP) of DYKDDDDK-tagged proteins from in vitro protein expression systems, bacteria, yeast, and mammalian cells. The amino acid sequence DYKDDDDK, commonly known as FLAG, is recognized by a high-affinity rat monoclonal antibody (clone L5) that is covalently attached to a magnetite-embedded agarose core particle.
- For protein purification, the magnetic agarose is added to a sample containing DYKDDDDK-tagged protein(s) with the tag on either the N- or the C-terminus. Captured proteins are then magnetically separated from the supernatant, and non-specifically bound proteins can be washed away before dissociating bound DYKDDDDK-tagged protein(s) with elution buffer. The magnetic agarose is removed from the solution using a magnetic stand or an instrument such as the KingFisher Flex Magnetic Particle Processor. Automated instruments are especially useful for higher throughput purifications and screening of purification conditions.
- Specific—Unique base-beads and highly specific antibody minimizes off-target binding (low non-specific binding)
- High purity —Optimized Bind-Wash-Elute protocol enables highpurity
- High Yield —special antibody conjugation method enables high yield
- Rapid—entire purification protocol typically takes less than 40 mins
- Economical - purification protocol allows multiple reuses
- Versatile—beads are compatible with manual and automated workflows (e.g., KingFisher instruments)
Specifications
Specifications
| Product Line | Pierce |
| Product Type | Magnetic Agarose |
| Quantity | 4 mL |
| Content And Storage | 1 mL settled beads. 4 mL supplied as a 25% suspension in PBS containing 0.02% sodium azide. Store at 2°C to 8°C |
| Format | Liquid |
| Stationary Phase | Anti-DYKDDDDK monoclonal antibodies |
| Shipping Condition | Wet Ice |
| Form | Liquid |
| Matrix | Magnetic Agarose |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction