Learn More
Thermo Scientific™ Pierce™ Anti-HA Agarose, 5 mL PROMO
Description
An HA tag is frequently incorporated into recombinant proteins for a variety of purposes. An anti-HA antibody can then be used to detect the protein when doing studies with transfected cells.
- Host Species: Mouse
- Cross Reactivity: Tag
- Isotype: IgG1
- Clone: 2-2.2.14
- Immunogen: HA peptide YPYDVPDYA derivitized to ovalbumin
- Formulation: 1 mL Purified Antibody in PBS, with 0.05% sodium azide; pH7.2
- Synonyms: human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
26181 is supplied as 1 mL of antibody on crosslinked 4% beaded agarose supplied as a 50% slurry in 0.1M phosphate, 0.15M NaCl, pH 7.2 with 0.05% sodium azide. Specific for the HA peptide YPYDVPDYA. The anti-HA agarose has been used successfully in immunoprecipitation applications. The anti-HA agarose has 3.5mg mouse anti-HA IgG1 monoclonal antibody conjugated permL of settled agarose resin. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.
Specifications
Specifications
| Product Type | Agarose |
| Column Type | Agarose Resin, Affinity |
| Format | Bottle |
| Conjugate | HA-epitope tag (YPYDVPDYA) |
| Purification Target | Anti-HA Antibody |
| Content And Storage | 5 mL of settle resin, provided as 10 mL of 50% slurry. Store at 2°C to 8°C |
| Quantity | 5 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.