Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Anti-HA Magnetic Beads, 1 mL PROMO

Catalog No. PI88836PM
Click to view available options
:
Thermo Scientific™ Pierce™ Anti-HA Magnetic Beads

Catalog No. PI88836PM

Supplier: Thermo Scientific™ 88836

Perform immunoprecipitation of recombinant HA-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.

The blocked magnetic bead surface is coated with anti–HA antibody, a highly specific mouse IgG1 monoclonal antibody that recognizes the HA-epitope tag (YPYDVPDYA) derived from the human influenza hemagglutinin (HA) protein. The Pierce Anti-HA Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms, such as the Thermo Scientific™ KingFisher Instruments, for high-throughput workflows.

Highlights:

  • Specific – highly specific anti–HA monoclonal antibody enables high–yield and high-purity immunoprecipitation (IP)

  • Fast – optimized protocol is completed in approximately 1 hour

  • Low nonspecific binding – stable, pre–blocked beads provide highly purified HA-tagged proteins

  • Versatile – beads are compatible with manual and automated workflows (e.g., Thermo Scientific™ KingFisher Instruments)

Specifications

Content And Storage Store at 4°C.
Form Liquid
Product Line Pierce
Quantity 1 mL
Protein Form Recombinant
Target HA
Type Magnetic Affinity Bead
Ligand Type Antibody

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.