Learn More
Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP PROMO
Description
The Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP provides biotinylation reagents designed specifically for the labeling of primary antibodies used in immunoprecipitation applications.
- Optimized for IP - reagents and protocols developed specifically to address antibody labeling for immunoprecipitation applications
- Easy to use - kit configuration specifically optimized to accommodate 50 to 200μg of antibody in a 100μL reaction; desalting columns are provided to remove excess biotinylation reagent after labeling
- Amine reactive - reacts with primary amines (-NH2) of antibodies
- Enhanced solubility - pegylation improves water solubility of the biotinylated antibody and prevents aggregation upon storage
- Reduced steric hindrance - longer spacer arm (29 angstroms) minimize steric hindrance when binding to avidin molecules
The kit contains sufficient reagents to label 50 to 200μg of antibody in 100μL reaction volumes for eight samples. The NHS-PEG4-biotin labeling reagent contains an amine-reactive N-hydroxysuccinimide ester (NHS) group and a water-soluble PEG4 spacer for optimal labeling and is provided in easy-to-use, single-use microtubes. Both the labeling efficiency of the biotinylation reagent and binding affinity of the labeled antibody have been validated using mouse monoclonal (IgG1, IgG2), rabbit polyconal, and rabbit monoclonal antibodies. Zeba™ Desalting Spin Columns are provided for easy and efficient removal of salts and excess biotin.
Applications:
- IP and co-IP
- IP and co-IP for mass spectrometry applications
- Antibody attachment to streptavidin agarose and magnetic supports
Specifications
Specifications
| Product Type | Antibody Biotinylation Kit |
| Quantity | 8 Reactions |
| Content And Storage | EZ-Link NHS-PEG4-Biotin, 8 x 0.5 mg, Zeba Spin Desalting Columns (7K MWCO), 8 x 0.5 mL, 20X Phosphate Buffered Saline, 15 mL Store in refrigerator 2°C to 8°C). |
| Labeling Scale | 100 μg |
| Product Line | Pierce |
| Label or Dye | Biotin |
| Labeling Target | Antibodies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.