Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP
GSA_VA

Catalog No. pi90407
Click to view available options
Quantity:
8 Reactions

Label primary antibodies with biotin for use in agarose or magnetic bead immunoprecipitation experiments using the Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP.

The Pierce Antibody Biotinylation Kit reagents have been optimized and validated to biotin label antibodies for IP and co-IP reactions. Determining the optimal number of biotins to attach to the target molecule is one of the major challenges of biotinylation. For IP and co-IP applications, too many biotins result in reduced affinity for the target antigen, while too few biotins result in antibody leaching upon elution of the target antigen. The biotin labeling procedure in the Pierce Antibody Biotinylation Kit for IP has been developed to address this challenge.

The Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP provides biotinylation reagents designed specifically for the labeling of primary antibodies used in immunoprecipitation applications.

  • Optimized for IP - reagents and protocols developed specifically to address antibody labeling for immunoprecipitation applications
  • Easy to use - kit configuration specifically optimized to accommodate 50 to 200μg of antibody in a 100μL reaction; desalting columns are provided to remove excess biotinylation reagent after labeling
  • Amine reactive - reacts with primary amines (-NH2) of antibodies
  • Enhanced solubility - pegylation improves water solubility of the biotinylated antibody and prevents aggregation upon storage
  • Reduced steric hindrance - longer spacer arm (29 angstroms) minimize steric hindrance when binding to avidin molecules

The kit contains sufficient reagents to label 50 to 200μg of antibody in 100μL reaction volumes for eight samples. The NHS-PEG4-biotin labeling reagent contains an amine-reactive N-hydroxysuccinimide ester (NHS) group and a water-soluble PEG4 spacer for optimal labeling and is provided in easy-to-use, single-use microtubes. Both the labeling efficiency of the biotinylation reagent and binding affinity of the labeled antibody have been validated using mouse monoclonal (IgG1, IgG2), rabbit polyconal, and rabbit monoclonal antibodies. Zeba™ Desalting Spin Columns are provided for easy and efficient removal of salts and excess biotin.

Applications:

  • IP and co-IP
  • IP and co-IP for mass spectrometry applications
  • Antibody attachment to streptavidin agarose and magnetic supports

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Biotinylation Kit
Quantity 8 Reactions
Content And Storage EZ-Link NHS-PEG4-Biotin, 8 x 0.5 mg
Zeba Spin Desalting Columns (7K MWCO), 8 x 0.5 mL
20X Phosphate Buffered Saline, 15 mL

Store in refrigerator (2–8°C).
Labeling Scale 100 μg
Product Line Pierce™
Label or Dye Biotin
Labeling Target Antibodies
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.