Learn More
Thermo Scientific™ Pierce™ Antibody Clean-up Kit PROMO
Description
This is a complete and easy-to-use kit for removing BSA and gelatin stabilizing proteins (up to 1%) from IgG samples. The protocol uses Thermo Scientific Melon™ Gel to bind these frequently used storage proteins, allowing the antibody to flow through in a purified form. The antibody is recovered in an amine-free and carboxyl-free buffer, ready for use in downstream applications such as biotinylation, fluorescent labeling, crosslinking, immobilization and enzymatic fragmentation (e.g., Fab preparation). Bovine serum albumin (BSA) and gelatin are often added to purified antibody solutions as stabilizers for long-term storage. Inclusion of 0.2 to 1% (2 to 10 mg/mL) BSA or gelatin helps stabilize dilute antibody solutions. Generally these added proteins do not interfere with immunodetection methods; however, their presence in solution does interfere with amine-reactive conjugation chemistries that are the basis for most biotinylation, fluorescent dye labeling, antibody fragmentation and covalent immobilization methods. For such methods, the antibody must be isolated from the stabilizing proteins, and the Antibody Clean-up Kit accomplishes this efficiently.
Highlights:
Fast – removes carrier protein from antibody solutions in half the time required for purifying antibodies through Protein A or G purification procedures
Gentle – no need to neutralize or desalt to mitigate the effects of harsh elution buffers
Complete – supplied desalting columns provide for easy buffer exchange into the required purification buffer or quick exchange into final buffer of choice
Economical – sufficient reagents to perform 10 antibody clean-up reactions
Includes:
Melon Gel IgG Purification Support and Buffer, desalting columns, microcentrifuge spin columns and tubes
Specifications
Specifications
| Product Type | Antibody Clean-up Kit |
| Product Line | Pierce |
| Column Type | Proprietary Resin, Affinity |
| Format | Kit, Microcentrifuge Spin Column |
| Stationary Phase | Proprietary Ligand |
| Purification Target | IgG Antibodies |
| Content And Storage | Sufficient For: Purifying 10 x 0.5 mL IgG samples, Melon Gel IgG Purification Support, 1.2 mL, Melon Gel Purification Buffer, 100 mL, Zeba Spin Desalting Columns, 7K MWCO, 2 mL, 10 Columns, Spin Columns, 0.8 mL, 10 Columns, Microcentrifuge Tubes, 2 mL, 30 Tubes, Store at 4°C |
| Quantity | 10 Reactions |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.