Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Microplate BCA Protein Assay Kit - Reducing Agent Compatible
GSA_VA

Catalog No. pi23252
Click to view available options
Quantity:
275 mL

Reducing agent-compatible version of our popular BCA Reagent Kit to measure protein concentration in samples containing DTT or BME thiol-reductants.

This reducing agent-compatible (RAC) version of the Thermo Scientific™ Pierce BCA Protein Assay provides accurate determination of protein concentration in samples that contain dithiothreitol (DTT), 2-mercaptoethanol (BME), TCEP and other disulfide reducing agents.

The BCA Protein Assay Kit - Reducing Agent Compatible (BCA-RAC) provides all of the advantages of the original BCA Assay, plus compatibility with disulfide reducing agents at concentrations routinely used in protein sample buffers. This special adaptation of the popular Pierce BCA Protein Assay method enables accurate protein concentration measurement for samples containing DTT, 2-ME or TCEP. Available in tube and microplate formats, these reducing agent compatible (RAC) BCA Kits extend the already broad reagent compatibility of the BCA Protein Assay protocol to include nearly all types of components commonly present in protein research samples.

Highlights:

  • Compatible – assay samples that contain up to 5mM DTT, 35mM BME, or 10mM TCEP

  • BCA technology – only a slight modification of the standard BCA Protein Assay protocol (15-minute incubation with Compatibility Reagent); no precipitation steps required

  • Small samples – requires only 25μL (standard kit) or less than 10μL (microplate kit) of sample

  • Colorimetric – measure with a standard spectrophotometer or plate reader (562nm)

  • Excellent uniformity – exhibits less protein-to-protein variation than dye-binding methods

  • High linearity – linear working range for BSA equals 125 to 2000μg/mL

Includes:

BCA Reagents A and B, Compatibility Reagent, Reconstitution Buffer, and Albumin Standard (2mg/mL); Microplate kit also includes assay-compatible 96-well microplates

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: 980 microplate assays with 9 μL samples
• BCA Reagent A, 250 mL
• BCA Reagent B, 25 mL
• Compatibility Reagent, 48 x 10 mg
• Reconstitution Buffer, 15 mL
• Albumin Standard Ampules, 2mg/mL, 10 x 1 mL
• 96-Well Microplates, 20 plates

Store at room temperature
Description Pierce BCA Microplate Protein Assay Kit, Reducing Agent Compatible (RAC)
Assay BCA Assay
Detection Method Colorimetric
Product Line Pierce™
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
Sufficient For 980 Microplate Assays
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 275 mL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.