Learn More
Thermo Scientific™ Pierce™ Protein A/G IgG Binding Buffer
Description
Thermo Scientific™ Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose beads.
The Gentle Ag/Ab Binding and Elution Buffer system uses near-neutral, high-salt conditions for elution, enabling even sensitive and labile antibodies (or other proteins) to be eluted from an affinity system without denaturation and inactivation. The phosphate-free binding buffer in this system is a necessary substitute for the Protein A binding buffer (pH 8), which precipitates with the Gentle Ag/Ab Elution Buffer.
Finally, the Mouse IgG1 Mild Elution Buffer, when used in combination with Protein A resin, enables selective elution and separation of the IgG1 subclass from total bound mouse IgG.
Highlights:
Three optimized binding buffers – for Protein A, Protein G or Protein A/G
Two elution buffer options – acidic (pH 2.8) or high salt (near-neutral)
Special system for mouse IgG1 – selective, two-stage elution from Protein A agarose
Item-Specific Features:
Protein A IgG Binding Buffer: pH 8.0; phosphate-based; contains glycerol and EDTA
Protein A/G IgG Binding Buffer: pH 8.0; phosphate-based; contains EDTA
Protein G IgG Binding Buffer: pH 5.0; contains 0.02% sodium azide
IgG Elution Buffer: pH 2.8; amine-based
Gentle Ag/Ab Binding Buffer: pH 8.0; phosphate-free; contains 0.05% sodium azide
Gentle Ag/Ab Elution Buffer: pH 6.6; high salt (ionic strength)
Mouse IgG1 Mild Elution Buffer: pH 6.0; contains 0.02% sodium azide
Includes:
Kits contain both binding and elution buffers
Specifications
Specifications
| Chemical Name or Material | Binding Buffer |
| Description | Pierce Protein A/G IgG Binding Buffer |
| pH | 8 |
| Physical Form | Liquid |
| Product Line | Pierce™ |
| Quantity | 240 mL |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.