Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Boronic Acid Resin PROMO

Catalog No. PI20244PM
Click to view available options
:
Thermo Scientific™ Pierce™ Boronic Acid Resin

Selectively bind and remove or purify ribonucleotides and small oligonucleotide RNAs using polyacrylamide beads of immobilized m-aminophenyl-boronic acid.

This immobilized boronic acid is an easy-to-use polyacrylamide affinity support for purification of ribonucleotides and other small molecular weight compounds that contain cis-diol groups. The ligand (m-aminophenyl-boronic acid) binds to the cis-diol groups on the sugar portion of nucleotides, forming a reversible five-member ring complex. After washing away non-bound molecules from the sample, the complex can be dissociated and the nucleotide eluted by lowering the pH or by addition of sorbitol.

Highlights:

  • Ligand – boronic acid

  • Support – beaded polyacrylamide (1800 MW exclusion limit)

  • Spacer – m-aminophenyl group

  • Loading – 100μmol boronate per milliliter of settled resin

  • Capacity – at least 99% binding and recovery of 110μmol AMP per milliliter of settled resin

  • Format – 50% resin slurry in 0.05% sodium azide

Specifications

Content And Storage Store at 4°C.
Form Resin Slurry
Product Line Pierce™
Quantity 10 mL
Target Ribonucleotide
Type Resin
Stationary Phase Boronic Acid
Format Bottle
Column Type Affinity, Polyacrylamide Resin

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.