Learn More
Thermo Scientific™ Pierce™ BSA Protein Digest, MS grade PROMO
Description
The Thermo Scientific Pierce BSA Protein Digest Standard, LC-MS Grade, is a validated protein digest optimized for use as a quality control sample for liquid chromatography (LC) and mass spectrometry (MS) analysis of proteomic samples.
- Positive control sample - BSA digest optimized and validated as a quality control standard for mass spectrometry applications
- Excellent sequence coverage - greater than 85% sequence coverage
- Validated peptide quality - digestion procedure optimized for minimal missed cleavages and over-alkylation
- Rigorously tested - high-quality, consistent BSA protein digest documented via lot-specific certificates of analysis
- Stable - provided in a stable lyophilized format
The Pierce BSA Protein Digest Standard is produced using high quality bovine serum albumin (BSA) and mass spectrometry-grade trypsin. The BSA is fully reduced, alkylated (iodoacetamide), and desalted (C18) after trypsin digestion to provide a quality LC/MS-grade standard that is free of intact protein. The peptide digest standard provides excellent sequence coverage with minimal over-alkylation and missed cleavages. This digest standard has been robustly tested for peptide quality, digestion efficiency, and lot-to-lot uniformity and is suitable for LC and LC-MS applications.
Specifications
Specifications
For Use With (Equipment) | Mass Spectrometer |
Product Type | Protein Digest |
Product Line | Pierce™ |
Content And Storage | Store in freezer (-5°C to -30°C). |
Starting Material | Peptides |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Calibration |
Quantity | 1 nmol |
Form | Solid |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.