Learn More
Thermo Scientific™ Pierce™ c-Myc-Tag IP/Co-IP Kit
Description
Thermo Scientific™ Pierce c-Myc Tag IP/Co-IP Kit provides the affinity resin and other reagents necessary to easily perform immunoprecipitation (IP) or co-immunoprecipitation (co-IP) experiments using a c-Myc-tagged protein as the bait.
Highlights:
Specific – the immobilized anti-c-Myc monoclonal antibody binds the c-Myc epitope tag with high specificity, providing high yield immunoprecipitation products and clean Western blot detection
High capacity – excellent results with as little as 1μg of anti-c-Myc antibody in IP mode with the positive control lysate
Rigorous – kit includes a positive control lysate that contains over-expressed GST-c-Myc to validate the reagents and specific protocol, and the instructions provide tips for planning other controls
Robust – the affinity system is compatible with IP or Co-IP from various cell lysates and physiological (non-denaturing) buffer systems; choose kits with or without mammalian cell lysis buffer
Convenient and easy – complete kit includes all necessary reagents, convenient spin columns, and easy-to-follow instructions
Includes:
M-PER™ Mammalian Protein Extraction Reagent (Part No. 23625 only), anti-c-Myc agarose resin, TBS pack, elution buffer, lane marker nonreducing sample buffer, positive control lysate, microcentrifuge spin columns and collection tubes

Specifications
Specifications
Description | Pierce c-Myc-Tag IP/Co-IP Kit |
Content And Storage | Sufficient For: 25 IP reactions, each using 5 μL resin (5 μg anti-c-Myc antibody) • Anti-c-Myc Agarose Resin, 250 μL • c-Myc-tagged Positive Control (Part No. 23633), 500 μL • TBS Pack (makes 500 mL), 1 pack • Elution Buffer (pH 2.0), 2 x 25 mL • Conditioning Buffer (100X), 5 mL • Non-Reducing Sample Buffer (5X), 5 mL • Spin Columns and Accessories, 27 columns • Microcentrifuge Collection Tubes, 100 tubes Store the c-Myc-tagged Positive Control at -80°C. Store Application Set at 4°C. |
Assay | Affinity Purification |
For Use With (Equipment) | Microcentrifuge |
Product Line | Pierce™ |
Quantity | 25 reactions kit |
For Use With (Application) | Pull-Down (Tag-Based), Immunoprecipitation (IP) |
Target | Target-Specific |
Type | Tag IP/Co-IP Kit |
Sufficient For | 25 Reactions |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.