Learn More
Thermo Scientific™ Pierce™ C18 Spin Columns, 25 Columns PROMO
Description
Pierce C18 Spin Columns
Thermo Scientific Pierce C18 Spin Columns are ready-to-use centrifuge columns of porous C18 reverse-phase resin for peptide sample preparation for MS and other analysis methods. The protocol has been optimized to efficiently remove interfering contaminants and release peptides in MS-compatible solutions. Columns can be used effectively for processing peptides derived from 10 ng to 30 μg of protein. Sensitivity and detection limits are dependent on the downstream application.
- Efficient—effectively removes interfering contaminants to improve spectral quality and is suitable for a variety of reverse-phase-compatible contaminants
- Flexible—works with a wide variety of load volumes and concentrations; no need to reduce sample volume before application
- Convenient—spin column format requires no special equipment to process multiple samples simultaneously
- Robust—special C18 resin allows excellent recovery percentages, even at low (sub-picomole) sample loads
Pierce C18 Spin Tips
Thermo Scientific Pierce C18 Spin Tips enable fast and efficient capture, concentration, desalting, and elution of up to 10 μg of peptides per 20 μL of sample with volumes ranging from 10 to 100 μL. The five-step procedure is simple and requires less than five minutes to process protein digests, strong cation exchange fractions, and other protein and peptide samples for mass spectrometric (MS) analyses. The Pierce C18 Spin Tips offer excellent flow properties with a high-efficiency monolithic C18 sorbent for fast wetting, loading, washing, and eluting.
Efficient—effectively removes interfering contaminants to improve spectral quality and is suitable for a variety of reverse-phase-compatible contaminants
- Rapid—C18 fast-flow tips have low resistance and improved flow
- Convenient—spin tips are provided with tip adaptor for easy centrifugation
- Cleaner sample—device filters particulates that can cause autosampler and column clogging
Applications
- Clean-up of peptides after protein digestion
- Peptide sample concentration and desalting
- Sample preparation for MALDI peptide analysis and proteomics workflows
Matrix-assisted laser desorption ionization (MALDI) and electrospray ionization (ESI) mass spectrometry (MS) are essential for studying biological compounds because of the high sensitivity and mass accuracy they provide. However, many of the buffers and compounds common to biological samples interfere with both MALDI-MS and ESI-MS, resulting in reduced sensitivity and poor-quality spectra. Pierce C-18 Spin Columns remove interfering contaminants and release peptides in MS-compatible solutions, providing an effective tool that aids in the MS examination of post-translational modifications or for the identification of proteins by peptide mapping.
Specifications
Specifications
For Use With (Equipment) | Microcentrifuge, Mass Spectrometer |
Product Type | Column |
Product Line | Pierce™ |
Content And Storage | Store at room temperature. |
Starting Material | Peptides, Protease-digested Protein |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Sample Clean Up |
Quantity | 25 Columns |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.