Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ CDI-activated Agarose Resin

Catalog No. PI20259 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20259 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20259 Supplier Thermo Scientific™ Supplier No. 20259
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Covalently immobilize N-nucleophiles and primary amine ligands in aqueous or organic solvent conditions using carbonyldiimidazole-activated resin.

Thermo Scientific™ Pierce CDI-Activated Agarose is a porous, beaded chromatography support that is activated with carbonyldiimidazole groups for covalent immobilization of amine-containing affinity ligands.

Pierce CDI-activated Agarose is a porous, beaded chromatography support that is activated with carbonyl diimidazole groups for covalent immobilization of proteins and other molecules that contain N-nucleophiles. Coupling can be performed in aqueous or organic solvent conditions, making possible the immobilization of many kinds of ligands. Once a molecule is immobilized, the prepared affinity resin can be used multiple times in typical affinity purification procedures.

Highlights:

  • Reliable coupling chemistry – immobilization occurs through the reaction of N-nucleophiles with 1,1'-carbonyldiimidazole groups of the resin to form stable, uncharged N-alkylcarbamate linkages

  • Easy to use – no secondary reagents needed; just wash equilibrate resin in alkaline coupling buffer and add ligand; reaction proceeds spontaneously

  • Stable activation – half-life of hydrolysis is much longer than hydroxysuccinimide ester activations, making immobilization reactions easier to prepare and control; simplifies filtration and washing before adding a ligand or protein

  • Well-defined coupling condition – reaction is most efficient with primary amines at pH 9 to 11

Properties of CDI-Activated Agarose:

  • Resin: crosslinked 6% beaded agarose
  • Activation: at least 50μmol 1,1'-carbonyldiimidazole (CDI) groups per milliliter of resin
  • Format: supplied as stabilized, 50% slurry in acetone
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Product Line Pierce
Quantity 10 mL
Target Primary Amine
Type Resin
Stationary Phase Carbonyl Diimidazole
Format Bottle
Column Type Affinity, Agarose Resin
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.