Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Centrifuge Columns, 5 mL, 25 Columns PROMO

Catalog No. PI89896PM
Click to view available options
:
Thermo Scientific™ Pierce™ Centrifuge Columns

Catalog No. PI89896PM

Supplier: Thermo Scientific™ 89896

Pack these empty 0.5 to 10 mL spin columns with agarose beads to perform small-scale affinity purification methods in 15- or 50-mL conical microcentrifuge tubes.

0.8 mL columns

  • Maximum volume: 800 μL
  • Recommended resin volume: 40–400 μL
  • Receiver tube: fits standard microcentrifuge tubes
  • Cap: O-ring screw-top
  • Twist-off bottom

2 mL columns

  • Maximum volume: 5 mL (2 mL resin bed, 3 mL sample)
  • Recommended resin volume: 400 μL–2 mL
  • Receiver tube: fits standard 15 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

5 mL columns

  • Maximum volume: 8 mL (5 mL resin bed, 3 mL sample)
  • Recommended resin volume: 1–5 mL
  • Receiver tube: fits standard 15 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

10 mL columns

  • Maximum volume: 22 mL (10 mL resin bed, 12 mL sample)
  • Recommended resin volume: 2–10 mL
  • Receiver tube: fits standard 50 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

20 mL columns

  • Maximum volume: 34 mL (20 mL resin bed, 14 mL sample)
  • Recommended resin volume: 4–20 mL
  • Receiver tube: fits standard 50 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

Applications

  • Affinity purification, affinity chromatography
  • Immuno-depletion
  • Ion exchange resins
  • Gel filtration, desalting, buffer exchange

Specifications

Bed Volume 2 mL
Capacity (Metric) 5 mL
Content And Storage Store at room temperature.
Product Line Pierce™
Quantity 25 Columns
Type Spin Column
Sample Volume 3 mL
Format Spin Column
Column Type Empty Column, Polyethylene Filter

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.