Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ Centrifuge Columns, 800 μL, 50 Columns PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ Centrifuge Columns
Description
0.8 mL columns
- Maximum volume: 800 μL
- Recommended resin volume: 40–400 μL
- Receiver tube: fits standard microcentrifuge tubes
- Cap: O-ring screw-top
- Twist-off bottom
2 mL columns
- Maximum volume: 5 mL (2 mL resin bed, 3 mL sample)
- Recommended resin volume: 400 μL–2 mL
- Receiver tube: fits standard 15 mL conical centrifuge tubes
- Cap: screw-top
- Twist-off bottom and press-on cap to reseal
5 mL columns
- Maximum volume: 8 mL (5 mL resin bed, 3 mL sample)
- Recommended resin volume: 1–5 mL
- Receiver tube: fits standard 15 mL conical centrifuge tubes
- Cap: screw-top
- Twist-off bottom and press-on cap to reseal
10 mL columns
- Maximum volume: 22 mL (10 mL resin bed, 12 mL sample)
- Recommended resin volume: 2–10 mL
- Receiver tube: fits standard 50 mL conical centrifuge tubes
- Cap: screw-top
- Twist-off bottom and press-on cap to reseal
20 mL columns
- Maximum volume: 34 mL (20 mL resin bed, 14 mL sample)
- Recommended resin volume: 4–20 mL
- Receiver tube: fits standard 50 mL conical centrifuge tubes
- Cap: screw-top
- Twist-off bottom and press-on cap to reseal
Applications
- Affinity purification, affinity chromatography
- Immuno-depletion
- Ion exchange resins
- Gel filtration, desalting, buffer exchange
Specifications
Specifications
Bed Volume | 400 μL |
Capacity (Metric) | 800 μL |
Content And Storage | Store at room temperature. |
Product Line | Pierce™ |
Quantity | 50 Columns |
Type | Spin Column |
Sample Volume | 800 μL |
Format | Microcentrifuge, Spin Column |
Column Type | Empty Column, Polyethylene Filter |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction