Learn More
Thermo Scientific™ Pierce™ Chicken IgY Purification Kit, 1 L PROMO
Description
The IgY Purification Kit is simple and easy to use. After isolating a yolk using the supplied egg separator, lipids are effectively removed by incubation and centrifugation in Pierce Delipidation Reagent. This first reagent is also effective as a storage solution, enabling multiple yolks to be collected and stored for simultaneous extraction at a later date. Upon addition of the specialized Pierce IgY Precipitation Reagent and subsequent centrifugation, a nearly pure pellet of intact IgY results; it is easily dissolved in phosphate buffered saline (PBS) or other storage buffer. The purified IgY is ready for use in immunodetection methods or antigen-specific purification without interference from lipids.
Highlights:
Cost-effective – purifies twice as much IgY per egg than other popular kits, providing a lower cost-per-milligram of purified IgY
High purity – method yields chicken antibody that is 85 to 95% pure (based on SDS-PAGE analysis)
Easy to use – simple precipitation method purifies chicken IgY without affinity columns
Flexible protocol – eggs can be harvested and quickly stored in buffer for purification at a later date
Convenient – use eggs directly out of refrigerator; no need to equilibrate them to room temperature
Includes:
Egg separator, delipidation reagent and IgY precipitation reagent
Specifications
Specifications
| Product Type | IgY Purification Kit |
| Product Line | Pierce™ |
| Column Type | Precipitation |
| Format | Kit, Bottle |
| Purification Target | IgY Antibody |
| Content And Storage | Sufficient For: Purifying chicken IgY from 5 egg yolks, Delipidation Reagent, 500 mL, IgY Precipitation Reagent, 500 mL, Egg Separator, 1 Separator, Store at 4°C. |
| Quantity | 1 L |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.