Learn More
Thermo Scientific™ Pierce™ Chloroacetamide, No-Weigh™ Format, 10 x 2 mg PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ Chloroacetamide, No-Weigh™ Format
Description
Alkylating reagents iodoacetamide and chloracetamide are sulfhydryl-reactive alkylating reagents used to block reduced cysteine residues for protein characterization and peptide mapping. Alkylation with iodoacetamide or chloracetamide after cysteine reduction results in the covalent addition of a carbamidomethyl group (57.07 Da) and prevents the formation of disulfide bonds. Reducing agents added after alkylation will react with excess iodoacetamide.
Specifications
Specifications
| Product Type | Alkylating Reagent |
| Quantity | 10 x 2 mg |
| Includes | Chloroacetamide |
| Product Line | Pierce |