Learn More
Thermo Scientific™ Pierce™ Streptavidin Coated Plate IP Kit
Description
Thermo Scientific™ Pierce Coated Plate IP Kits enable immunoprecipitation experiments to be performed in 96-well microplates.
Highlights:
- Ready-to-use, high quality coated plates provide high capacity and consistency
- Accommodate antibody and sample volumes comparable to tube-based methods
- Easily recover immunoprecipitated samples for downstream analysis
- Coated plates are 96-well strip plates, convenient for experiments requiring only a partial plate
- Include simple instructions, including detailed explanation of appropriate controls
- Two kits available, suitable for most common antibody types (mouse, rabbit, human and goat IgG subclasses) or any biotinylated antibody or bait protein
Advantages of the Plate Format:
- Best suited for simultaneously processing multiple samples and their control conditions
- Allows faster, easier and more thorough washing than with traditional tube/resin IP methods
- Makes use of familiar and convenient ELISA tools (multichannel pipettors and plate washing)
- No no tubes to open and close, and no centrifugation required
Protein A/G Kit (Part No. 45350)
- Binds all human IgG subclasses; human IgM, IgA (weak), and IgE; and all mouse IgG subclasses
- Offers increased binding for mouse monoclonal antibodies compared to Protein A or Protein G alone
- Free of pH dependency when binding IgGs, providing more flexible binding conditions
Streptavidin Kit (Part No. 45360)
- Used in combination with biotinylated antibodies, resulting in IP and co-IP preparations free of interfering antibody fragments
- Contains biotin-blocking buffer to minimize endogenous biotin binding
Includes:
Coated 96-well strip plates, uncoated 96-well strip plates, PBS pack, detergent solution, elution buffer, neutralization buffer and sealing tape
Recommended for:
Rapid, trouble-free immunoprecipitation of multiple samples and their controls; Immunoprecipitation when plate-based tools and techniques are preferred
Specifications
Specifications
| Description | Pierce Streptavidin Coated Plate IP Kit |
| Content And Storage | Sufficient For: Assaying 48 samples with controls or 192 total IP reactions • Streptavidin Coated 96-well Strip Plates, 2 plates • Uncoated 96-well Strip Plate (white), 2 plates • PBS Pack (makes 500 mL), 1 pack • Biotin Blocking Buffer, 2 x 15 mL • Surfact-Amps X-100 Detergent Solution, 6 x 10 mL • Elution Buffer (pH 2.8), 50 mL • Neutralization Buffer, 12 mL • Sealing Tape for 96-well Plates, 18 sheets Store at 4°C. |
| Assay | Affinity Purification |
| For Use With (Equipment) | Microplate Mixer |
| Product Line | Pierce |
| Quantity | 1 Kit |
| For Use With (Application) | Immunoprecipitation (IP), Pull-Down (Biotin-Based) |
| Target | Not Target-Specific |
| Type | Immunoprecipitation/Co-Immunoprecipitation Kit |
| Sufficient For | 192 samples |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.