Learn More
Thermo Scientific™ Pierce™ Conjugate Purification Kit PROMO
Description
The metal chelate ligand on the column binds the enzyme conjugates while allowing free enzyme to pass through.
Highlights:
- Reduced background for blotting and ELISA
- Fewer washes needed to remove background
- Increased sensitivity of assay system with more active reagent
- Improved signal:noise ratio of conjugate
Includes:
Immobilized Iminodiacetic acid 1 x 2 mL column, Polyacrylamide Desalting Column 1 x 10 mL column, FreeZyme Column Activator Buffer 25 mL, FreeZyme Elution Buffer 35 mL, BupH™ Tris Buffered Saline Pack two each, BupH Phosphate Buffered Saline Pack one each, Activity Preservation Solution 1 mL
Specifications
Specifications
| Product Type | Conjugate Purification Kit |
| Quantity | 1 Kit |
| Sufficient For | 10 purifications, each of 0.5 to 50 mg of enzyme-conjugated antibody |
| Content And Storage | Sufficient For: 10 purifications, each of 0.5 to 50 mg of enzyme-conjugated antibody, IDA Agarose Column, 2 mL, 1 column, Polyacrylamide Desalting Column, 10 mL, 1 column, Column Activator Buffer, 25 mL, Elution Buffer, 35 mL, TBS Packs (each makes 500 mL), 2 packs, PBS Pack (makes 500 mL), 1 pack, Activity Preservation Solution, 1 mL Store Kit at 4°C. |
| Labeling Scale | 0.5 to 3 mg |
| Product Line | Pierce™ |
| Label or Dye | HRP (Horseradish Peroxidase) |
| Labeling Target | IgG |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.