Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ Crossflow Diafiltration Reservoir, 500 mL PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ Crossflow Diafiltration Reservoir
Description
- For continuous buffer replacement using Pierce Cross Flow Filtration Cassettes.
Specifications
Specifications
| Color | Clear |
| Material | Plastic |
| Sterility | Non-sterile |
| Type | Self-standing Reagent Reservoir with Lid |
| Includes | 3 ports |
| Quantity | 1 Ea. |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| Bottom Shape | Flat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction