Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Crosslink IP Kit
GSA_VA

Catalog No. PI26147 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26147 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26147 Supplier Thermo Scientific™ Supplier No. 26147
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Reagent kit and protocol for crosslinking IP antibodies to Protein A/G agarose to enable antigen immunoprecipitation without antibody contamination.

Thermo Scientific™ Pierce Crosslink IP Kit is an adaptation of traditional immunoprecipitation protocols that enables antigen recovery without antibody contamination or co-elution.

This immunoprecipitation kit extends the functionality of traditional immunoprecipitation (IP) methods by adding crosslinking technology and microcentrifuge spin column sample handling to the procedure. The primary benefits resulting from these features are the ability to purify target protein without contamination by the antibody and the ability to more effectively wash and separate samples from the beaded agarose resin.

The Pierce Crosslink IP Kit method involves capturing the IP antibody to Protein A/G Agarose resin and covalently immobilizing it to the support by crosslinking with disuccinmidyl suberate (DSS). The antibody resin is then incubated with the sample that contains the protein antigen of interest, allowing the antibody:antigen complex to form. After washing to remove nonbound (presumably undesired) components of the sample, the antigen is recovered by dissociation from the antibody with elution buffer supplied in the kit. The entire procedure is performed in a microcentrifuge spin cup, allowing solutions to be fully separated from the agarose resin upon brief centrifugation. Only antigen is eluted by the procedure, enabling it to be identified and further analyzed without interference from antibody fragments. Furthermore, the antibody resin often can be reused for additional rounds of immunoprecipitation.

Highlights:

  • Eliminates antibody contamination – antibody is irreversibly attached to the agarose beads so that co-elution of heavy and light chains with the purified protein is minimized

  • Easy and efficient scalability – use only the amount of antibody needed for a single IP experiment (e.g., 5μg) or immobilize up to 200μg of antibody to prepare a larger quantity of ready-made IP affinity resin for many experiments

  • Assay reliability and sample handling – Pierce Spin Columns eliminate resin loss and provide for more efficient separation of solutions than traditional IP methods that use only microcentrifuge tubes

  • Prepared antibody affinity resin is reusable – because the antibody is covalently immobilized and usually is not inactivated by the mild elution procedure, the resin often can be used several times

  • Complete kits – package includes sufficient reagents and spin columns to perform at least 50 antibody immobilizations and IP experiments

Includes:

Pierce Protein A/G Plus Agarose, control agarose resin, DSS crosslinker, coupling buffer, IP lysis/wash buffer, conditioning buffer, Tris-buffered saline, elution buffer, sample loading buffer, spin columns and collection tubes

Recommended for:

Immunoprecipitation followed by electrophoresis and excision of the protein for mass spectrometry or sequencing identification analysis; Immunoprecipitation and SDS-PAGE analysis of a target protein whose molecular weight is similar to the heavy or light chain antibody fragment; Immunoprecipitation of target proteins for subsequent analysis by nondenaturing methods (i.e., methods not involving SDS-PAGE)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: Preparing and performing at least 50 IP reactions, each with 10 μL of resin
• Protein A/G Plus Agarose, 0.55 mL
• DSS Crosslinker, 8 x 2 mg
• Coupling Buffer (20X), 25 mL
• IP Lysis/Wash Buffer, 2 x 50 mL
• Conditioning Buffer (100X), 5 mL
• Tris-Buffered Saline (20X), 25 mL
• Elution Buffer, 50 mL
• Lane Marker Sample Buffer (5X), 5 mL
• Control Agarose Resin, 2 mL
• Spin Columns and Collection Tubes

Store at 4°C. Store DSS desiccated at 4°C.
Assay Affinity Purification
For Use With (Equipment) Microcentrifuge
Product Line Pierce
Quantity 50 reactions
For Use With (Application) Immunoprecipitation (IP)
Target Not Target-Specific
Type Immunoprecipitation/Co-Immunoprecipitation Kit
Sufficient For 50 Reactions
Format Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.