Learn More
Thermo Scientific™ Pierce™ Cysteine-HCl PROMO
Description
Thermo Scientific™;Pierce Cysteine-HCl is purified, crystalline cysteine hydrochloride salt (MW 175.6) that is useful as a sulfhydryl standard in assay with Ellman's Reagent, or as an additive in protein refolding experiments.
Specifications
Specifications
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 25°C. |
| Product Line | Pierce |
| Form | Solid |
| Quantity | 5 g |
| Product Type | Cysteine Hydrochloride |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.