Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ D-Luciferin, Monopotassium Salt
Click to view available options
Quantity:
100 mg
1 g
Salt Form:
Monopotassium Salt
Monosodium Salt
Description
Thermo Scientific™ Pierce D-Luciferin is synthesized and manufactured to ensure high performance at a low cost, and the reporter is also tested by HPLC to ensure proper chirality and activity.
We are the only supplier to perform chiral-high performance liquid chromatography (HPLC) testing to ensure that the product is not only pure but also in the active form. Our D-Luciferin outshines the competition on many levels.
Specifications
Specifications
| Content And Storage | Store at or below -20°C. |
| Detection Method | Bioluminescence |
| For Use With (Application) | ATP Assay, Gene Reporter Assay |
| Product Type | D-Luciferin, Monosodium Salt |
| Product Line | Pierce™ |
| Quantity | 100 mg |
| Salt Form | Monosodium Salt |
| Substrate Properties | Chemical Substrate |
| Substrate Type | Luciferase Substrate |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction