Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Dextran Desalting Columns, 5K MWCO, 10 mL, 5 Columns PROMO

Catalog No. PI43233PM
Click to view available options
:
Thermo Scientific™ Pierce™ Dextran Desalting Columns

Ready-to-use, gel-filtration Thermo Scientific Pierce Dextran Desalting Columns can separate proteins and macromolecules from low-molecular weight buffer salts and reagents up to 5000 Daltons (5 kDa) in 5- or 10-mL volumes.

Applications

  • Removing salts from protein solutions
  • Eliminating phenol from nucleic acid preparations
  • Separating excess crosslinker from conjugate preparations
  • Removing excess derivatizing agents from modified proteins
  • Withdrawing unreacted dye from fluorescent antibodies
  • Eliminating free radiolabel from labeled proteins
  • Buffer exchange

Gravity-flow gel filtration using dextran desalting columns

Gravity-flow gel filtration involves the chromatographic separation of molecules of different dimensions based on their relative abilities to penetrate a suitable stationary phase. A chromatographic matrix, usually consisting of very small, uncharged porous particles in an aqueous solution, is packed into a column and then used for separation via size exclusion. Different levels of separation can be achieved based on the pore size of the medium packed into the desalting column—in our case, dextran. With size exclusion filtration, large molecules are excluded from the internal pores of the gel and emerge from the column first. Smaller molecules can penetrate the pores, then progress through the column at a slower rate. These smaller molecules are subsequently flushed through the column with additional buffer volume. Our Dextran Desalting Columns are rigid for easy handling and provide excellent flow properties. Dextran is stable in water, salt solutions, organic solvents and alkaline or weakly acidic solutions. Dextran is also heat-stable and can be autoclaved dry or in solution at a neutral pH for 30 minutes at 120°C without affecting its chromatographic properties.

Specifications

Product Type Desalting Column
Content And Storage Store at room temperature or 4°C.
Format Gravity Flow Column
Purification Target Buffer Exchange, Protein
Column Type Size-exclusion, Dextran Resin
MWCO 5.0 kDa
Product Line Pierce™
Disposable Single-use
Quantity 5 Columns
Volume (Metric) 10 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.