Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Direct IP Kit

Catalog No. PI26148 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26148 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26148 Supplier Thermo Scientific™ Supplier No. 26148
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Immobilize IP antibodies (any species or class) on agarose beads without Protein A/G to immunoprecipitate antigens without antibody interference.

Thermo Scientific™ Pierce Direct IP Kit enables immunoprecipitation with any species or class of antibody and yields purified antigen without contamination from the IP antibody.

Additional Information and Online Ordering

This direct immunoprecipitation kit represents a significant advancement in immunoprecipitation (IP) technology as it replaces the use of immobilized Protein A or Protein G with a method for direct covalent attachment of antibodies to the beaded agarose resin. The primary benefits resulting from this method are the opportunity to use any species or subclass of purified antibody (not just types that bind to Protein A or G) and the ability to purify target protein without contamination by the antibody. The method also makes it possible to immunoprecipitate antigens from serum samples without co-purifying non-target immunoglobulins. Finally, the kit uses microcentrifuge spin cups to effectively wash and separate samples from the beaded agarose resin.

Highlights:

  • Requires less than 10μg of antibody – optimized procedure requires only 2 to 10μg of antibody for a single IP experiment, no more than is required for traditional IP methods

  • Easy and efficient scalability – use only the amount of antibody needed for a single immunoprecipitation or immobilize up to 200μg of antibody to prepare a larger quantity of ready-made IP affinity resin for many experiments

  • Minimal antibody contamination – antibody is irreversibly attached to the agarose beads so that co-elution of heavy and light chains with the purified protein is minimized (i.e., less than 5%)

  • IP with any species and subclass of antibody – Use chicken IgY, human IgE, mouse IgM or any other purified protein

  • Prepared antibody affinity resin is reusable – because the antibody is covalently immobilized and not inactivated by the mild elution procedure, the resin often can be used several times

  • Convenient sample handling – Pierce Spin Columns eliminate resin loss and provide for efficient separation of solutions

  • Complete kit – package includes cell lysis buffer and sufficient reagents and spin columns to perform at least 50 antibody immobilizations and IP experiments

  • Requires purified antibody – IP antibody solution must be free of BSA, gelatin or other stabilizer proteins; for easy removal of these proteins, see Part No. 44600

Direct immunoprecipitation in two easy steps:

Step 1: Attach antibody to agarose resin

The first step in the Direct IP Method is to prepare the IP antibody affinity resin. Pure antibody is incubated in phosphate buffer with an appropriate volume of AminoLink™ Plus Coupling Resin, an aldehyde-activated beaded agarose resin. During the incubation, various lysine epsilon-amines on the surface of the large antibody molecule react with aldehyde groups on the resin to form Schiff-base bonds that are then stabilized to secondary amine bonds in the presence of cyanoborohydride. The immobilization reaction, known as reductive amination, is nondenaturing and typically results in greater than 85% antibody coupling efficiency with minimal loss of antigen-binding function.

Step 2: Perform immunoprecipitation experiments

Once prepared, the antibody resin can be used in several parallel aliquots or in total for IP experiments. Antibody resin is incubated with a sample that contains the protein antigen of interest, allowing the antibody:antigen complex to form. After washing to remove nonbound (presumably undesired) components of the sample, the antigen is recovered by dissociation from the antibody with elution buffer supplied in the kit. The entire procedure is performed in a microcentrifuge spin cup, allowing solutions to be fully separated from the agarose resin upon brief centrifugation. Only antigen is eluted by the procedure, enabling it to be identified and further analyzed without interference from antibody fragments. Furthermore, the antibody resin can be reused for additional rounds of immunoprecipitation.

Includes:

AminoLink Plus Coupling Resin, coupling buffer, quenching buffer, wash solution, sodium cyanoborohydride solution, IP lysis/wash buffer, conditioning buffer, Tris-buffered saline, elution buffer, lane marker sample buffer, control agarose resin, spin columns and collection tubes

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: Preparing and performing at least 50 IP reactions, each with 10 μL of resin
• AminoLink Plus Coupling Resin, 2 mL
• Coupling Buffer (20X), 25 mL
• Quenching Buffer, 50 mL
• Wash Solution, 50 mL
• Sodium Cyanoborohydride Solution (5M), 0.5 mL
• IP Lysis/Wash Buffer, 2 x 50 mL
• Conditioning Buffer (100X), 5 mL
• Tris-Buffered Saline (20X), 25 mL
• Elution Buffer, 50 mL
• Lane Marker Sample Buffer (5X), 5 mL
• Control Agarose Resin, 2 mL
• Spin Columns and Collection Tubes

Store at 4°C.
Assay Affinity Purification
For Use With (Equipment) Microcentrifuge
Product Line Pierce
Quantity 50 reactions
For Use With (Application) Pull-Down, Immunoprecipitation (IP)
Target Antibody
Type Immunoprecipitation/Co-Immunoprecipitation Kit
Sufficient For 50 Reactions
Format Kit
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.