Learn More
Thermo Scientific™ Pierce™ Disposable Columns, 10 mL PROMO
Description
These bulk packages of empty columns are economical and convenient devices for packing affinity purification supports and desalting resins for gravity-flow procedures. Three columns sizes are offered, suitable for procedures requiring 0.5 to 10 mL resin-bed volumes. The plastic columns are supplied with matching pairs of polyethylene discs and polypropylene caps (top and bottom). The columns are economically-priced for single use and sufficiently durable to prepare and store columns for multiple uses.
Highlights:
- For bench top gravity-flow affinity purification and size exclusion chromatography
- Supplied complete with porous polyethylene discs, stoppers and end caps
- Columns can be pre-packed and stored until needed
- Compatible with most types of aqueous buffer eluents commonly used in chromatography
- Plastic funnels can be attached to expand the buffer reservoir for increased flow rates
Includes:
Trial pack contains two columns of each available size (six columns total), matching column accessories (discs and caps), and one funnel (column reservoir extender)
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Content And Storage | Store at room temperature. |
| Product Line | Pierce™ |
| Quantity | 100 Columns |
| Type | Spin Column |
| Format | Gravity Flow Column |
| Column Type | Empty Column, Polyethylene Filter |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.