Learn More
Thermo Scientific™ DNA Coating Solution
Description
Perform efficient DNA surface coating of microplates and microsample tubes using Thermo Scientific™ Pierce DNA Coating Solution.
Highlights:
- No more overnight incubations or heating microplates and waiting
- Promotes efficient immobilization (by adsorption) of DNA in microplate wells (polystyrene)
- Coated target DNA permits speed and flexibility in probing by hybridization and other means
- DNA coated onto plastic surfaces is stabilized sufficiently to allow storage for months before use
- Solution has been autoclaved to inactivate DNase
Specifications
Specifications
| Buffer | Immunoassay Buffer, Hybridization Buffer |
| Content And Storage | Store at room temperature. |
| Description | Pierce DNA Coating Solution |
| Formulation | Proprietary buffered solution |
| Product Type | DNA Coating Solution |
| Quantity | 100 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.