Learn More
Description
Thermo Scientific™ Pierce DSP Crosslinker (Lomant's Reagent) is an amine-reactive crosslinker with NHS-ester reactive ends and a cleavable disulfide bond in its spacer arm.
Highlights:
- Cleavable by reducing agents
- Membrane-permeable
- Acylate in aqueous or organic media within two minutes at room temperature
- Four- to five-hour half-life of ester at pH 7.0
- Reactive groups: NHS ester (homobifunctional)
- Reactive toward: amino groups
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Chemical Reactivity | Amine-Amine |
| Cleavable | By Thiols |
| Description | DSP |
| Molecular Weight (g/mol) | 404.42 |
| PEGylated | No |
| Spacer Arm Length | 12.0 Å |
| Content And Storage | Upon receipt store desiccated at 4°C. |
| Cell Permeability | Yes |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |
Frequently Asked Questions (FAQs)
DSP (dithiobis(succinimidyl propionate)), Lomant's Reagent is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
DSP has been estimated to be soluble in acetonitrile up to a concentration around 25 mg/mL.
Yes, frequently the resultant reaction mixture will become hazy after the addition of the stock solution.
DSP has been used for, but is not limited to, the following applications: examining spatial relationships of capsid polypeptides, studying renal Na+ and K+ ATPase, nearest neighbor relationships of bovine mitochondrial H+ATPase, chemical cross-linking of a-CPI, identifying cross-linked cytochrome P-450 in rat liver microsomes, studying the influence of metal ions on prothrombin self-association, studying the glycoprotein topology on intact human red blood cells, and determining membrane antigens by covalent cross-linking to monoclonal antibodies.
A homobifunctional cross-linker has the same reactive group on both ends, for example two amine-reactive groups. It is used in a one-step conjugation where the two molecules are mixed prior to the addition of the cross-linker. This leads to less efficient cross-linking because polymers can easily be created. A heterobifunctional cross-linker has two different reactive groups, for example an amine-reactive group on one end and a sulhydryl reactive group on the other. These can be used in two-step conjugations by reacting the fIrst end with the molecule containing its reactive group, and then adding the molecule that is reactive towards the other end. This leads to much more efficient cross-linking because polymerization does not occur.
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
